Search Thermo Fisher Scientific
Catalog Number | Volume (Metric) | Product Type |
---|---|---|
FKS10785126 | 20 mL | Luer-slip Syringe |
FIS03-377-20 | 1 mL | Luer-slip Syringe |
FKS10280655 | 1 mL | Luer-slip Syringe |
FIS03-377-23 | 10 mL | Luer-slip Syringe |
FKS10178894 | 10 mL | Luer-slip Syringe |
FIS03-377-25 | 30 mL | Luer-slip Syringe |
FIS03-377-22 | 5 mL | Luer-slip Syringe |
FKS10271245 | 5 mL | Luer-slip Syringe |
FIS03-377-29 | 10 mL | Luer-lock Syringe |
Dispense and filter small volumes of liquids with these disposable syringes, used with Thermo Scientific™ Titan3™ or Thermo Scientific™ Target2™ syringe filters. Thermo Scientific™ National Target All-Plastic Disposable Syringes are an ideal choice for all syringe filter applications. These disposable syringes feature two-part construction using polypropylene barrels and polyethylene plungers. This eliminates the need for rubber or synthetic plunger gaskets, and requires no silicone or oil lubricant in the barrel.