Nalgene™ Unwire™ Test Tube Racks: Resmer™ Manufacturing Technology
Nalgene™ Unwire™ Test Tube Racks: Resmer™ Manufacturing Technology
Actual product may vary
Nalgene™ Unwire™ Test Tube Racks: Resmer™ Manufacturing Technology
Nalgene™ Unwire™ Test Tube Racks: Resmer™ Manufacturing Technology
Nalgene™ Unwire™ Test Tube Racks: Resmer™ Manufacturing Technology
Fisher Catalogue
Nalgene™ Unwire™ Test Tube Racks: Resmer™ Manufacturing Technology
Securely hold a full load of test tubes or centrifuge tubes for 13, 16, 20, 25 and 30mm diameter sizes with these strong, durable racks.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberColorArrayFor Use With (Equipment)
FKS10778721Blue3 x 830mm Tubes
5970-0513
also known as GT5970-0513TC
Red6 x 1213mm Tubes
FIS14-809-60Red6 x 1213mm Tubes
5970-0213Yellow6 x 1213mm Tubes
5970-0113Orange6 x 1213mm Tubes
5970-0013White6 x 1213mm Tubes
FIS14-809-22White6 x 1213mm Tubes
FIS14-809-62Red6 x 1216mm Tubes
FKS10435071Red6 x 1216mm Tubes
5970-0316
also known as NLG5970-0316
Blue6 x 1216mm Tubes
FIS14-809-43Blue6 x 1216mm Tubes
FKS10098210Blue6 x 1216mm Tubes
5970-0416
also known as NLG5970-0416
Green6 x 1216mm Tubes
FKS10343461Yellow6 x 1216mm Tubes
5970-0116
also known as NLG5970-0116
Orange6 x 1216mm Tubes
FIS14-809-24White6 x 1216mm Tubes
5970-0520
also known as GT5970-0520TC
Red4 x 1020mm Tubes
FIS14-809-64Red4 x 1020mm Tubes
5970-0320Blue4 x 1020mm Tubes
FIS14-809-52Blue4 x 1020mm Tubes
FIS14-809-38Yellow4 x 1020mm Tubes
5970-0020White4 x 1020mm Tubes
FIS14-809-26White4 x 1020mm Tubes
FKS10291761White4 x 1020mm Tubes
FIS14-809-66Red4 x 1025mm Tubes
FKS10663721Red4 x 1025mm Tubes
5970-0425
also known as NLG5970-0425
Green4 x 1025mm Tubes
5970-0530
also known as 5970-0530TC
Red3 x 830mm Tubes
FIS14-809-68Red3 x 830mm Tubes
5970-0330Blue3 x 830mm Tubes
FIS14-809-44Blue3 x 830mm Tubes
5970-0430Green3 x 830mm Tubes
FIS14-809-49Green3 x 830mm Tubes
5970-0230Yellow3 x 830mm Tubes
FIS14-809-50Yellow3 x 830mm Tubes
FIS14-809-36Orange3 x 830mm Tubes
5970-0030White3 x 830mm Tubes
FIS14-809-30White3 x 830mm Tubes
FKS10644101White3 x 830mm Tubes
Catalog number FKS10778721
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Color:
Blue
Array:
3 x 8
For Use With (Equipment):
30mm Tubes
Securely hold a full load of test tubes or centrifuge tubes for 13, 16, 20, 25 and 30mm diameter sizes with Thermo Scientific™ Nalgene™ Unwire™ Test Tube Racks. Molded in a single piece for exceptional strength and durability, these racks have no coating to chip, peel or rust.
  • Smooth finish leaves fewer places for contaminants to lodge; no welds to break, no sharp edges.
  • Endplates accept large labels for easy identification of samples.
  • Higher specific gravity than water so it stays submerged in a water bath and won't float.
  • Withstands most lab chemicals and temperatures up to 121°C for a variety of lab procedures.
  • Available in six bright colors to help keep samples organized.
  • 15mL conical tubes fit 20mm racks; 50mL conical fit 30mm racks.
  • No assembly is required; rack is ready to use.
Please recycle the corrugated cartons and LDPE bags in which your Nalgene products are packaged.
Specifications
Product TypeTest Tube Rack
DescriptionTest Tube Rack Unwire™ Blue 30mm
Array3 x 8
AutoclavableAutoclavable
ColorBlue
HoldsTubes (30 mm)
MaterialRESMER
Positions24
ShapeRectangular
Shelves3 Tiers
For Use With (Equipment)30mm Tubes
Dimensions (L x W x H)11 x 4.25 x 3.62 in. (28.3 x 10.8 x 9.2 cm)
Length (Metric)28.3 cm
Length (English)11 in.
Width (Metric)10.8 cm
Width (English)4.25 in.
Height (Metric)9.2 cm
Height (English)3.62 in.
Diameter (Metric) Opening30 mm
Diameter (Metric) Inner30 mm
Unit SizeEA