Conductivity Standard, NIST Traceable, 1,000μS/cm, Reagecon™
Conductivity Standard, NIST Traceable, 1,000μS/cm, Reagecon™
Fisherbrand
Conductivity Standard, NIST Traceable, 1,000μS/cm, Reagecon™
Accredited to ISO 17025. Reagecon™ Conductivity Standard, NIST Traceable, 1,000μS/cm is an aqueous-based, non-hazardous standard with a high degree of accuracy, low temperature variation and guaranteed stability through the expiration date, even after opening the bottle.
Have Questions?
Catalog NumberQuantity
FKS10778448500 mL
Catalog number FKS10778448
Price (SGD)
565.00
EA
Add to cart
Quantity:
500 mL
Request bulk or custom format
Price (SGD)
565.00
EA
Add to cart
Specifications
Shelf Life18 months
Certificate of AnalysisYes
Conductivity1,000μS/cm (±1% at 25°C)
Physical FormLiquid
For Use With (Equipment)Conductivity Meter
View more
This conductivity standard is aqueous based, thereby eliminating any errors attributable to matrix mismatch. Aqueous standards are non-hazardous for easy storage, handling and disposal. Each bottle is supplied with a comprehensive Certificate of Analysis and has a table of conductivity variation with temperature printed on the label.
  • Convenience and lower costs—Reduced health, safety and environmental requirements and paperwork
  • Very low temperature coefficient of variation—Reduces measurement errors and enables non-temperature controlled conditions or field work
  • Guaranteed stability through expiry date—Eliminates the need to open a fresh bottle of standard every time it is used
  • High accuracy (±1% at 25°C) —Enables it to be used as a calibrator or control, in accordance with the most exacting requirements (e.g. USP)