Mixer Mill Grinding Balls
Mixer Mill Grinding Balls
Actual product may vary
Fisherbrand
Mixer Mill Grinding Balls
Stainless steel grinding balls for use in mixer mills
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDiameter (English)
FKS107778210.78 in.
FKS104587220.47 in.
Catalog number FKS10777821
Price (SGD)
103.00
EA
Add to cart
Diameter (English):
0.78 in.
Price (SGD)
103.00
EA
Add to cart
  • Composition is approximately 84.5% Fe, 13% Cr.
  • Approximate density is 7700kg/m3
Specifications
Product TypeGrinding Ball
Composition84.5% Fe, 13% Cr
Diameter (English)0.78 in.
Diameter (Metric)2 cm
For Use With (Application)Ideal for increasing the degree of mixing, grinding
For Use With (Equipment)MM 200, 301 mixer mill, S 100 centrifugal ball mill and PM 400 planetary ball mill