1x1 image pixel for data collection
Porcelain Buchner Funnel
Porcelain Buchner Funnel
Actual product may vary
Fisherbrand
Porcelain Buchner Funnel
Designed with glazed porcelain for excellent thermal and chemical resistance. Haldenwanger™ Porcelain Buchner Funnels are an excellent choice for quick filtration.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Diameter (Metric) Plate
FKS10771752135 mL74mm
FKS10179630580 mL114mm
Catalog number FKS10771752
Price (SGD)
292.00
EA
Add to cart
Capacity (Metric):
135 mL
Diameter (Metric) Plate:
74mm
Price (SGD)
292.00
EA
Add to cart
Designed with glazed porcelain for excellent thermal and chemical resistance. Haldenwanger™ Porcelain Buchner Funnels are an excellent choice for quick filtration.
  • Vertical sides
  • Available in a variety of sizes to fit many filtration needs
  • Perforated fixed filter plate
Specifications
Capacity (Metric)135 mL
MaterialPorcelain
Diameter (Metric) Plate74mm
Quantity1Pack
Product TypeBuchner Funnel
Unit SizeEA
Documents & Downloads
Certificates
Safety Data Sheets
Share catalog number, name or link