Borosilicate Glass Short Neck Round Bottom Flask
Actual product may vary
Fisherbrand
Borosilicate Glass Short Neck Round Bottom Flask
Use these flasks for a variety of applications. Quickfit™ Borosilicate Glass Short Neck Round Bottom Flask are manufactured from borosilicate glass for corrosion-free performance and resistance to thermal shock.
Have Questions?
Catalog NumberCapacity (Metric)Socket Joint
FKS1076511125 mL14/23
Catalog number FKS10765111
Price (SGD)
348.00
Pack
Capacity (Metric):
25 mL
Socket Joint:
14/23
Price (SGD)
348.00
Pack
Use these flasks for a variety of applications. Quickfit™ Borosilicate Glass Short Neck Round Bottom Flask are manufactured from borosilicate glass for corrosion-free performance and resistance to thermal shock.
  • Hgh quality Quickfit™ ground glass socket
  • Comprehensive range of 46 flasks from 5mL to 10,000mL
  • Body diameter and wall thickness comply with relevant ISO 1773 and ISO 4797 requirements
Specifications
Certifications/ComplianceISO 1773, ISO 4797
Capacity (Metric)25 mL
MaterialBorosilicate Glass
Height (Metric)85 mm
Neck StyleShort
Socket Joint14/23
Bottom ShapeRound
Unit SizePack