Secador™ Standard Desiccator Cabinet, Horizontal Model 1.0
Secador™ Standard Desiccator Cabinet, Horizontal Model 1.0
Actual product may vary
Fisherbrand
Secador™ Standard Desiccator Cabinet, Horizontal Model 1.0
Constructed of Durastar™ co-polyester. Standard desiccator cabinets block 99% of UV light and are resistant to staining, crazing, and chemical attack.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberColor
FKS10760583Clear with Blue Ends
FIS08-647-850Amber
Catalog number FKS10760583
Price (SGD)
1,306.00
EA
Add to cart
Color:
Clear with Blue Ends
Price (SGD)
1,306.00
EA
Add to cart
  • Amber color model reduces visible light penetration by over 50% making it the perfect choice for storing light sensitive materials like reagents, analytical standards and investigational compounds
  • Large doors maximize access to interior space and have a patented seal design, tab latches and security loops for a padlock or tamper-evident seals
  • Perforated shelves facilitate circulation of dry air; space for desiccant under bottom shelf
  • Easy-to-view digital hygrometer is located in the door front; side swinging door opens to the left
  • Stack up to 3 units high to save space
  • Can be used with desiccant cartridges, F420450250, F420450200, F420480100
Specifications
AutoclavableNot Autoclavable
ColorClear with Blue Ends
Cabinet StyleVertical
MaterialCopolyester
Product TypeDesiccator
Door StyleLeft-hinged and gasketed
For Use With (Application)Easy and reliable storage of moisture and light sensitive products, blocks 99% UV light, used with disposable or reusable desiccant 420450250
IncludesCarrying handles, one latch and loops for a padlock or tamper-evident seals, door-mounted hygrometer
Model1.0
No. of Shelves1
ResistanceChemicals, stains and fine cracks
Shelf TypeFixed Perforated
Volume (English)0.75 cu. ft.
Dimensions (L x W)16.3 x 13.4 in. (41.4 x 34.1 cm)
Length (English)16.3 in.
Length (Metric)41.4 cm
Width (English)13.4 in.
Width (English) Door8.9 in.
Width (Metric)34.1 cm
Width (Metric) Door22.5 cm
Height (English)8.4 in.
Height (English) Opening4.5 in.
Height (Metric)21.3 cm
Depth (English)16.3 in.
Depth (Metric)41.4 cm
Unit SizeEA