Check Order Status
PVC Pipette Tray
Actual product may vary
Fisherbrand
PVC Pipette Tray
Accommodates pipets of different lengths and diameters. Kartell™ PVC Pipette Tray is constructed with PVC for strength and durability.
Have Questions?
Catalog Number
Dimensions (L x W x H)
FKS10757541
300 x 426 x 30 mm
Catalog number
FKS10757541
Price (SGD)
125.00
EA
Add to cart
Dimensions (L x W x H):
300 x 426 x 30 mm
Price (SGD)
125.00
EA
Add to cart
Trays can be used as workstation or drawer organizer for pipets
Work station model has vertical and horizontal slots for individual placement of pipets
Longitudinal: 7 pipets with overall stem diameter not exceeding 20mm
Lateral: 16 pipets with stem diameter not exceeding 10mm
Drawer model has 4 compartments
Holds approximately 30 pipets
Accommodates pipets of 1, 2, 5 and 10mL capacities
Specifications
Capacity (Metric)
10 mL
Dimensions (L x W x H)
300 x 426 x 30 mm
Color
White
Length (Metric)
300 mm
Material
PVC
Type
Pipette Tray
Width (Metric)
426 mm
For Use With
Drawer
No. per Pack
1/Pk.
Shape
Rectangular