Sedimentation Imhoff Cones
Sedimentation Imhoff Cones
Actual product may vary
Fisherbrand
Sedimentation Imhoff Cones
Manufactured with SAN for crystal clear visibility and strength. Kartell™ Sedimentation Imhoff Cone is an ideal solution for determining settleable solids in surface and saline waters and sewage.
Have Questions?
Catalog NumberVolume (Metric)
FKS107561921000 mL
Catalog number FKS10756192
Price (SGD)
144.00
EA
Add to cart
Volume (Metric):
1000 mL
Price (SGD)
144.00
EA
Add to cart
Manufactured with SAN for crystal clear visibility and strength. Kartell™ Sedimentation Imhoff Cone is an ideal solution for determining settleable solids in surface and saline waters and sewage.
  • Permanently molded graduations provide an accurate measurement of mixtures
  • Polypropylene screw plug in base allows easy drainage
  • Back flush cleaning easily effected
Specifications
Volume (Metric)1000 mL
GraduatedYes
MaterialSAN
TypeSedimentation Imhoff Cone
Height (Metric)480 mm
Diameter (Metric) OuterTop: 125 mm, Base: 7.8 ± 0.2 mm
Unit SizeEA