Check Order Status
Whatman™ Lead Acetate Test Paper Dispenser
Actual product may vary
Fisherbrand
Whatman™ Lead Acetate Test Paper Dispenser
Dispenser for rapid qualitative test paper to detect hydrogen sulfide
Have Questions?
Catalog Number
Quantity
FKS10754925
Each
Catalog number
FKS10754925
Price (SGD)
106.00
PK
Add to cart
Quantity:
Each
Price (SGD)
106.00
PK
Add to cart
Dispenser for rapid qualitative test paper to detect hydrogen sulfide
Lead Acetate Test Paper Detections
As little as 5ppm of atmospheric or gas stream H
2
S
As little as 4ppm H
2
0
2
by preblackening paper with H
2
S
Specifications
For Use With (Application)
For rapid qualitative test paper to detect hydrogen sulfide, detect ∼5ppm of H2S in atmosphere or gas stream
Quantity
Each
Type
Lead Acetate Test Paper Dispenser