1x1 image pixel for data collection
Snap-together Conical Tube Rack
Snap-together Conical Tube Rack
Actual product may vary
Fisherbrand
Snap-together Conical Tube Rack
Keep conical tubes standing upright. Fisherbrand™ Snap-together Conical Tube Rack is designed for stable use in water baths but can also be used on benchtops.
Have Questions?
Catalog NumberHolds
FKS10753257Tubes (30 x 15 mL, 20 x 50 mL)
Catalog number FKS10753257
Price (SGD)
111.00
EA
Add to cart
Holds:
Tubes (30 x 15 mL, 20 x 50 mL)
Price (SGD)
111.00
EA
Add to cart
  • Use 15 and 50 mL tubes simultaneously
  • Shipped flat, easy-to-assemble
  • Alphanumeric grid reference aids for tube identification
  • Tubes:
    • 15 mL: 30 Wells, 17 mm dia.
    • 50 mL: 20 Wells, 28 mm dia.
  • Dimensions: 171 x 170 x 61 mm
  • Not recommended for autoclaving
Specifications
AutoclavableNot Autoclavable
ColorOrange
HoldsTubes (30 x 15 mL, 20 x 50 mL)
MaterialABS
Positions50
For Use With (Equipment)15 and 50mL conical tubes
Quantity1 Each
Dimensions (L x W x H)171 x 170 x 61mm
Length (Metric)171 mm
Width (Metric)170 mm
Height (Metric)61 mm
Diameter (Metric) Well17 and 28 mm
Unit SizeEA
Share catalog number, name or link