Chemical Resistant Plastic PVC Apron
Chemical Resistant Plastic PVC Apron
Actual product may vary
Fisherbrand
Chemical Resistant Plastic PVC Apron
Manufactured of polyvinyl chloride. BRAND™ Chemical Resistant PVC Apron includes eyelets, neck band and waist band welded into the apron. Reinforced fabric and construction is industry quality.
Have Questions?
Catalog number FKS10748134
Price (SGD)
184.00
EA
Add to cart
Price (SGD)
184.00
EA
Add to cart
Plastic apron made of PVC with eyelets, neck band and waist band welded into the apron, industry quality, black, fabric reinforced, size 80cm x 100cm. PVC
Specifications
TypeApron
MaterialPVC
ColorBlack
ConstructionReinforced plastic fabric
ResistanceChemicals
IncludesEyelets, neck band and waist band
Dimensions (L x W)80 x 100 cm
Length (Metric)80 cm
Width (Metric)100 cm
Unit SizeEA