DuraCal™ 2.00 Value pH Buffer, Hamilton™
Product Image
Fisherbrand
DuraCal™ 2.00 Value pH Buffer, Hamilton™
Economical choice since only about 20mL of buffer is used per calibration. Hamilton™ DuraCal™ 2.00 Value pH Buffers have actual values determined by a DKD laboratory, accredited for pH measurement.
Have Questions?
Catalog NumberQuantity
FKS10747444500 mL
Catalog number FKS10747444
Price (SGD)
135.00
EA
Add to cart
Quantity:
500 mL
Request bulk or custom format
Price (SGD)
135.00
EA
Add to cart
Specifications
pH2.00
Quantity500 mL
Unit SizeEA
  • 2.00 pH value
  • Immune to micro-organisms
  • Expiration date on the bottle
  • First class certificate with traceability to international standards