MF-Millipore™ Membrane Filter, 0.22 μm
MF-Millipore™ Membrane Filter, 0.22 μm
Actual product may vary
Fisher Catalogue
MF-Millipore™ Membrane Filter, 0.22 μm
MF-Millipore™ Membrane Filter is a 13 mm diameter, mixed cellulose esters (MCE) membrane, hydrophilic.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDiameter (Metric)
FKS1074291125 mm
FKS1059651247 mm
Catalog number FKS10742911
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Diameter (Metric):
25 mm
Designed for a wide range of analytical and research applications. MF-Millipore™ Mixed Cellulose Ester Membranes, 0.22μm pore size are available in a range of diameters. Composed of plain, white mixed cellulose membrane.
Biologically inert mixtures of cellulose acetate and cellulose nitrate have made MF-Millipore™ membrane filters one of the most widely used membranes in analytical and research applications.
MF-Millipore™ filters without Triton™ surfactant contain minimum amounts of wetting agent and have a lower water extractable content than standard MF-Millipore™ filters.
Features and Benefits:
  • Versatile filter for biological and environmental monitoring applications
  • Available in a range of pore sizes, colored black or white, with or without a gridded surface
  • Compatible with ethylene oxide, gamma irradiation, and autoclave sterilization methods
Specifications
AutoclavableNot Autoclavable
Bubble Point≥3.52 bar, air with water
ColorWhite
DisposableReusable
Filter TypeMembrane Filter
FormatDisc
MaterialMCE
Pore Size0.22 μm
SterilityNon-sterile
Flow Rate18 mL/min. x cm2
Gas Flow Rate2 L/min./cm2
HydrophobicityHydrophilic
Membrane TypePlain
No. per Case100 Pk.
Quantity100 Pk.
Thickness (Metric)150 μm
TypeFilter Membranes
WettabilityHydrophilic
Diameter (English)1 in.
Diameter (Metric)25 mm
Unit SizePK