BRANDplates™ pureGrade™ 96-Well Microplates
BRAND<i>plates</i>&trade; pureGrade&trade; 96-Well Microplates
Actual product may vary
Fisherbrand
BRANDplates™ pureGrade™ 96-Well Microplates
Constructed of polystyrene with nontreated surface. BRANDplates™ pureGrade™ 96-Well Microplates are noncytotoxic, and free from endotoxins, DNase, DNA, and RNase. Medium binding, nonsterile microplates are suited for homogenous assays, screening, and storage with gray alphanumeric embossing on white and black solid plates for easy well identification.
Have Questions?
Catalog NumberColorWell ShapeVolume (Metric) WellSurface Treatment
FKS10742694BlackFlat Bottom (F-well)350 μLNon-treated
Catalog number FKS10742694
Price (SGD)
1,267.00
PK
Add to cart
Color:
Black
Well Shape:
Flat Bottom (F-well)
Volume (Metric) Well:
350 μL
Surface Treatment:
Non-treated
Price (SGD)
1,267.00
PK
Add to cart
  • Polystyrene surface
  • Medium binding, nonsterile
  • Standard plate for most applications
  • Gray alphanumeric embossing on white and black solid plates for easy well identification
  • Strip plates available
  • Standard plates available in U-, V-, F-, C-shapes
  • Transparent bottoms available in F- shape
  • Available in clear, white or black
  • Free from endotoxins, DNase, DNA, RNase; noncytotoxic
  • Suited for homogenous assays, screening, and storage.
Specifications
BarcodeIncludes Barcode
Certifications/ComplianceANSI, SLAS
FormatStandard
LidWithout Lid
Array8 x 12
Binding TypeMedium Binding
ColorBlack
MaterialPolystyrene
Volume (Metric) Well350 μL
No. of Wells96
SterilityNon-sterile
Surface TreatmentNon-treated
Well ShapeFlat Bottom (F-well)
Dnase FreeYes
For Use With (Application)Homogenous assays, screenings and storage
Quantity100/Cs.
RNase FreeYes
Unit SizePK