Soda Lime Glass Culture Tubes with Screw Cap
Soda Lime Glass Culture Tubes with Screw Cap
Actual product may vary
Fisherbrand
Soda Lime Glass Culture Tubes with Screw Cap
Manufactured from soda lime glass. BRAND™ Soda Lime Glass Culture Tubes with Screw Cap include a polypropylene screw cap with white elastomer seal and GL thread. Autoclavable at 121°C.
Have Questions?
Catalog NumberLength (Metric)Thread Size
FKS10742512100 mmGL 18
Catalog number FKS10742512
Price (SGD)
378.00
PK
Add to cart
Length (Metric):
100 mm
Thread Size:
GL 18
Price (SGD)
378.00
PK
Add to cart
  • Manufactured from soda lime glass.
  • Includes polypropylene screw cap with white elastomer seal and GL thread.
  • Autoclavable (121°C).
Specifications
Closure MaterialPolypropylene
Length (Metric)100 mm
MaterialSoda Lime Glass
Quantity100/Pk.
Thread SizeGL 18
Diameter (Metric)16 mm
Closure TypeScrew Cap
Product TypeCulture Tube
Unit SizePK