Isopore™ Membrane Filters, 0.2 μm
Isopore™ Membrane Filters, 0.2 μm
Actual product may vary
Fisherbrand
Isopore™ Membrane Filters, 0.2 μm
Use for analyses of airborne contaminants and other particles using optical or electron microscopy. Isopore™ Polycarbonate Membrane Filters have a smooth, glass-like surface for clearer sample observation. Available in a variety of pore sizes/flow rates, diameters and colors.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDiameter (Metric)Color
FKS1074238147 mmBlack
FISGTTP0250025 mmWhite
Catalog number FKS10742381
Price (SGD)
2,641.00
PK
Add to cart
Diameter (Metric):
47 mm
Color:
Black
Price (SGD)
2,641.00
PK
Add to cart
Use for analyses of airborne contaminants and other particles using optical or electron microscopy. Isopore™ Polycarbonate Membrane Filters have a smooth, glass-like surface for clearer sample observation. Available in a variety of pore sizes/flow rates, diameters and colors.
The Isopore™ membrane is a polycarbonate, track-etched screen filter recommended for all analyses in which the sample is viewed on the surface of the membrane. Isopore™ membrane offers distinct advantages for the analysis of airborne contaminants and other particles using optical or electron microscopy. The Isopore™ membrane is composed of polycarbonate film, which has a smooth, glass-like surface for clearer sample observation. The unique manufacturing process of the membrane ensures a precise pore diameter and a consistent pore size for accurate separation of samples by size. Matched-weight filters are not usually required because of low, constant tar and ash weights.
Features & Benefits:
•Membrane structure retains particles on the surface, simplifying counting and analysis
•Isopore™ membranes do not stain, resulting in low background interference
•Non-hygroscopic, allowing for rapid drying and reduced sample analysis time
•Translucent material does not require clearing for transmitted light microscopy; also available in brown variety
Applications:
Air Monitoring, Epifluorescent Microscopy, Chemotaxis Assays
Specifications
AutoclavableNot Autoclavable
Bubble Point50 psi
ColorBlack
Filter TypeScreen Filter
MaterialPolycarbonate
Pore Size0.22 μm
SterilityNon-sterile
Flow RateAvg. 16 mL/min x cm2 (typical results @ 10 psi);6 mL/min x cm2/psi
For Use With (Application)Air Analysis; Cell Biology; Cell Culture; Bioassays; Cytology; Environmental Analysis; Epifluorescence.
Gas Flow Rate1.0 L/min/cm2
HydrophobicityHydrophilic
No. per Case100Pack
Quantity100/Pk.
TypeFilter Membranes
WettabilityHydrophilic
Diameter (Metric)47 mm
Unit SizePK