Check Order Status
18/10 Stainless Steel Micro Scoops, Half-rounded
Actual product may vary
Fisher Catalogue
18/10 Stainless Steel Micro Scoops, Half-rounded
Add Bochem™ 8/10 Stainless Steel Micro Scoops, Half-rounded to your arsenal of laboratory instruments.
Have Questions?
Change view
Catalog Number
Length (Metric)
Length (Metric) Blade
FKS10742352
185 mm
50 mm
FKS10350821
130 mm
40 mm
2 Options
Catalog number
FKS10742352
View Price:
Sign In
Sign in to see your account pricing. Need an account? Register with us today.
Length (Metric):
185 mm
Length (Metric) Blade:
50 mm
Type 1
Half-rounded
Also available in wide scoop
18/10 stainless steel
Specifications
Quantity
1 Ea.
Autoclavable
Autoclavable
Shape
Half-rounded
Product Type
Micro Scoop
Length (Metric) Blade
50 mm
Width (Metric)
4 mm
Width (Metric) Blade
4 mm
Blade Material
Stainless Steel (18/10)
Diameter (Metric) Outer
2mm
Dimensions (L x W)
185 x 4 mm
Material
Stainless Steel (18/10)
Length (Metric)
185 mm