DURAN™ HPLC Screw Cap, 4 Tubing Connectors, Blue, GL 45, PP
DURAN™ HPLC Screw Cap, 4 Tubing Connectors, Blue, GL 45, PP
Actual product may vary
Fisherbrand
DURAN™ HPLC Screw Cap, 4 Tubing Connectors, Blue, GL 45, PP
Screw cap for pressure and vacuum resistant bottle for HPLC applications.
Have Questions?
Catalog NumberThread Size (Metric)
FKS1074083412 mm
Catalog number FKS10740834
Price (SGD)
293.00
PK
Add to cart
Thread Size (Metric):
12 mm
Price (SGD)
293.00
PK
Add to cart
The DURAN™ HPLC bottle is available as a ready-made system. This comprises a special pressure and vacuum resistant bottle and a screw cap with 4 ports. This enables 3 stations to be served at the same time from one vessel. At the same time a sterile pressure equalisation can be fitted to the fourth outlet.
  • Connection of different hose diameters (1.6 and 3.2 mm)
  • Unused ports can be sealed with silicone blanking seals
  • Optional sterile pressure equalisation sets (membrane filter 0.2 μm)
  • Complete system comprising: DURAN™ pressure plus laboratory bottle with 4-port screw cap (PP); four screw connections (black, M8 thread) and silicone seals available
Specifications
DescriptionScrew cap HPLC, GL 45, 4 ports, complete (GL 45 screw cap, 4 x M8 black screw caps, 12 x silicone seals for 1.6; 3.2 mm tubing or blanks)
For Use With (Application)Safe transfer of liquid media within a closed and sterile system; Supply of solvents to HPLC equipment
MaterialPolypropylene, Silicone
Quantity2/Pk.
Thread Size (Metric)12 mm
Neck Finish12 mm