Polypropylene Amber Wide Neck Round Bottles
Polypropylene Amber Wide Neck Round Bottles
Actual product may vary
Fisherbrand
Polypropylene Amber Wide Neck Round Bottles
Store a wide range of light sensitive materials in these wide-neck plastic bottles. Azlon™ Polypropylene Amber Wide Neck Round Bottles feature a leak-proof, liner-less design for easy filling and emptying of solid or liquid samples.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)
FKS10739185150 mL
FKS10554925500 mL
Catalog number FKS10739185
Price (SGD)
80.00
PK
Add to cart
Capacity (Metric):
150 mL
Price (SGD)
80.00
PK
Add to cart
Store a wide range of light sensitive materials in these wide-neck plastic bottles. AzlonTM Polypropylene Amber Wide Neck Round Bottles feature a leak-proof, liner-less design and offer good chemical resistance.
  • Amber high quality, heavy duty bottles
  • Non-stick polypropylene screw caps
  • Good chemical resistance
  • Opaque, rigid amber polypropylene bottles with black polypropylene screw caps
  • Smooth inner contours for efficient pouring and ease of cleaning
  • Heavy-duty and very durable in use
  • Autoclavable at 121°C
Specifications
Capacity (Metric)150 mL
Capacity (English)5.07 oz.
ColorAmber
Diameter (Metric) Inner Neck32 mm
For Use With (Application)Ideal for a wide range of light sensitive liquid or solid storage
MaterialPolypropylene
Quantity10/PK
Closure ColorBlack
Certifications/ComplianceCCD 077; SW-CDC-001501; EN-8-1007-22-0912
Closure IncludedYes
AutoclavableAutoclavable
MouthWide
ShapeRound
Height (Metric)116 mm
Product TypeRound Wide Mouth Bottle
Height (English)4.56 in.
Closure MaterialPolypropylene
Diameter (Metric)51 mm
Closure TypeScrew Cap
Unit SizePK