Centrifuge Tubes with CentriStar™ Cap
Centrifuge Tubes with CentriStar™ Cap
Actual product may vary
Fisherbrand
Centrifuge Tubes with CentriStar™ Cap
Features CentriStar™ caps with an easy-on/easy-off flat top and offers advanced ergonomics with wider knurls and roll-over edge design for easier gripping. Corning™ Centrifuge Tubes with CentriStar™ Caps leak-proof design comes with a revolutionary plug feature that virtually eliminates all seepage when used under recommended conditions.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)GraduationsPackagingMax. RCF
FKS1073877115 mL1.5 to 14.5 mL in 0.5 mL intervalsSleeve12,500 x g
FIS05-526B50 mL5 to 50 mL in 2.5 mL incrementsRack17,000 x g
FKS1033413150 mL5 to 50 mL in 2.5 mL incrementsRack17,000 x g
FIS05-538-59B15 mL1.5 to 14.5 mL in 0.5 mL intervalsSleeve12,500 x g
FIS07-203-51050 mL5 to 50 mL in 2.5 mL incrementsUniversal Rack15,500 x g
Catalog number FKS10738771
Price (SGD)
872.00
PK
Add to cart
Capacity (Metric):
15 mL
Graduations:
1.5 to 14.5 mL in 0.5 mL intervals
Packaging:
Sleeve
Max. RCF:
12,500 x g
Price (SGD)
872.00
PK
Add to cart
Leak-proof design comes with a revolutionary plug feature that virtually eliminates all seepage when used under recommended conditions. Features CentriStar™ caps with an easy-on/easy-off flat top; offers advanced ergonomics with wider knurls and roll-over edge design for easier gripping.
 Superior Performance
  • Temperature range from -80°C to 120°C
  • Conical bottoms
  • Manufactured from clear virgin medical grade polypropylene (PP) for excellent visibility and strength
  • Sterile, certified RNase-/DNase-free and nonpyrogenic
  • Large white marking spot and black printed graduations
  • 5 year shelf life
Specifications
Certifications/ComplianceRNase, DNase-free
Closure MaterialHDPE
Closure ColorOrange
Diameter (English) Outside0.7 in.
ColorClear
Capacity (Metric)15 mL
DisposableSingle-use
MaterialPolypropylene
No. per Case500
No. per Pack50
PackagingSleeve
SterilitySterile
ShapeConical
Length (Metric)118 mm
Capacity (English)0.507 oz.
Bottom ShapeConical
Closure TypeCentriStar
Closure IncludedYes
Writing AreaLarge White Marking Area
Diameter (Metric)17.5 mm
Graduations1.5 to 14.5 mL in 0.5 mL intervals
Product TypeCentrifuge Tube
Max. RCF12,500 x g
Unit SizePK