Nalgene™ Square PETG Media Bottles with Closure: Sterile, Shrink-Wrapped Trays
Nalgene™ Square PETG Media Bottles with Closure: Sterile, Shrink-Wrapped Trays
Actual product may vary
Nalgene™ Square PETG Media Bottles with Closure: Sterile, Shrink-Wrapped Trays
Nalgene™ Square PETG Media Bottles with Closure: Sterile, Shrink-Wrapped Trays
Nalgene™ Square PETG Media Bottles with Closure: Sterile, Shrink-Wrapped Trays
Nalgene™ Square PETG Media Bottles with Closure: Sterile, Shrink-Wrapped Trays
Fisherbrand
Nalgene™ Square PETG Media Bottles with Closure: Sterile, Shrink-Wrapped Trays
Eliminate costly washing, depyrogenation and autoclaving steps with certified sterile media bottles, ideal for prolonged storage of pH sensitive liquids.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Closure Size
FKS10736923500 mL38-430 mm
342020-003030 mL20-415 mm
FKS1022300330 mL20-415 mm
342020-006060 mL24-415 mm
FKS1017693260 mL24-415 mm
342020-0125125 mL38-430 mm
342020-0250250 mL38-430 mm
FIS09-923-16E250 mL38-430 mm
FKS10531845250 mL38-430 mm
342020-0500500 mL38-430 mm
342020-2000
also known as 342020-200 0
2000 mL53B
FKS100690122000 mL53B
342020-1000
also known as 342020-100 0
1 L38-430 mm
FIS09-923-16C1 L38-430 mm
FKS103503431 L38-430 mm
Catalog number FKS10736923
Price (SGD)
1,061.00
PK
Add to cart
Capacity (Metric):
500 mL
Closure Size:
38-430 mm
Price (SGD)
1,061.00
PK
Add to cart
Eliminate costly washing, depyrogenation and autoclaving steps with certified sterile Thermo Scientific™ Nalgene™ Square PETG Media Bottles with Closure, ideal for prolonged storage of pH sensitive liquids such as culture media.
Features:
  • Supported by extensive validation documentation
  • Made from durable, break-resistant polyethylene terephthalate copolyester, glycol modified (PETG)2/O2
  • Molded in graduations and neck ring for use with tamper evident shrink sleeves
  • Non-pyrogenic and non-cytotoxic
  • Transparency and reduced permeability to CO2 and O2
  • Bottle and closure material are Animal Derived Component (ADC) Free
  • Leakproof†
Note:
2L size has molded-in handgrips.
† The term “leakproof” applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. B) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.
Specifications
Capacity (Metric)500 mL
Capacity (English)16.9 oz.
ColorClear
Description500mL, 38-430 closure, tray pack
Diameter (English)2.9 in.
Diameter (English) Inner Neck1.1 in.
Diameter (Metric) Inner Neck28 mm
For Use With (Application)Prolonged Storage
MaterialPETG
No. per Case40
No. per Pack20
SterilitySterile
Certifications/ComplianceBottles and closures are certified radiation sterilized (SAL 10-6), nonpyrogenic and noncytotoxic, and comply with USP Class VI guidelines.
Closure IncludedYes
AutoclavableNot Autoclavable
GraduatedYes
MouthNarrow
ShapeSquare
Fit for PurposeSterile
Height (Metric)177 mm
Product TypeMedia Bottle
Height (English)6.97 in.
Closure MaterialHDPE
Closure Size38-430 mm
Diameter (Metric)74 mm
Unit SizePK