Used for AKTA chromatography systems. Cytiva Polypropylene On-line Filter Kits are resistant to organic solvents and salt buffers commonly used in chromatography of biomolecules.
Have Questions?
Catalog Number
Temperature (Metric) Max
FKS10720740
4°C
Catalog number FKS10720740
Price (SGD)
-
Temperature (Metric) Max:
4°C
Kit contains 10 x filter 2 x Support screen
The wetted parts are resistant to organic solvents and salt buffers commonly used in chromatography of biomolecules, except 100 % Ethyl acetate, 100 % Hexane and 100 % Tetrahydrofuran