Stainless Steel Test Sieves, 200 Dia. x 50 mm H, Pore sizes in Micrometers, ISO 3310/1
Stainless Steel Test Sieves, 200 Dia. x 50 mm H, Pore sizes in Micrometers, ISO 3310/1
Actual product may vary
Fisherbrand
Stainless Steel Test Sieves, 200 Dia. x 50 mm H, Pore sizes in Micrometers, ISO 3310/1
Use for separation, fractioning, or particle size determination. These Stainless Steel test sieves have an internal height of 50 mm and diameter of 200 mm. Available in a wide range of pore sizes smaller than 1mm.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberPore Size
FKS10719132850 μm
FKS10166121300 μm
FKS1027916175 μm
FKS10269021500 μm
Catalog number FKS10719132
Price (SGD)
410.00
EA
Add to cart
Pore Size:
850 μm
Price (SGD)
410.00
EA
Add to cart
  • Stainless Steel frame with wire gauze
  • Diameter: 200 mm
  • Height: 50 mm
  • ISO 3310/1
Specifications
Quantity1 Each
Pore Size850 μm
TypeTest Sieve
Certifications/ComplianceISO 3310/1. Main Series R’10 according to DIN 323.
IncludesCertificate of compliance with ISO 3310/1 or NFX 11-504
Height (Metric)50mm
For Use With (Application)For separation, fractioning, particle size determination
Diameter (Metric) Outer200 mm
Frame MaterialStainless Steel
Mesh MaterialStainless Steel
Unit SizeEA