Polycarbonate Erlenmeyer Flasks
Polycarbonate Erlenmeyer Flasks
Actual product may vary
Fisherbrand
Polycarbonate Erlenmeyer Flasks
Manufactured from optically clear polycarbonate for optimum visibility and durability. Corning™ Disposable Polycarbonate Erlenmeyer Flasks are ideal for shaker cultures applications.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Diameter (Metric) Outer NeckClosure TypeGraduations
FKS10716851250 mL3.1 cmFlat25 mL
FIS07-200-668250 mL3.1 cmScrew Cap, Vented25 mL
FKS10022611250 mL3.1 cmScrew Cap, Vented25 mL
FIS07-200-669500 mL4.3 cmScrew Cap, Vented50 mL
FIS07-200-6721 L4.3 cmScrew Cap, Vented50 mL
FKS101141011 L4.3 cmScrew Cap, Vented50 mL
FIS07-200-670125 mL2.6 cmScrew Cap, Vented25 mL
FKS10045411125 mL2.6 cmScrew Cap, Vented25 mL
CRG430421125 mL2.6 cmFlat25 mL
FIS10-041-8125 mL2.6 cmFlat25 mL
FKS10654541125 mL2.6 cmFlat25 mL
CRG430183250 mL3.1 cmFlat25 mL
FIS10-041-18250 mL3.1 cmFlat25 mL
CRG430422500 mL4.3 cmFlat50 mL
FIS10-041-8B500 mL4.3 cmFlat50 mL
FKS10664541500 mL4.3 cmFlat50 mL
Catalog number FKS10716851
Price (SGD)
2,310.00
PK
Add to cart
Capacity (Metric):
250 mL
Diameter (Metric) Outer Neck:
3.1 cm
Closure Type:
Flat
Graduations:
25 mL
Price (SGD)
2,310.00
PK
Add to cart
  • Made from optically clear polycarbonate
  • Ideal for shaker culture applications
  • Available with flat caps or vented caps for applications requiring sterile gas exchange
  • Sterilized by gamma radiation and certified nonpyrogenic
  • RNase- and DNase-free
Disclaimer: This product is not intended to mitigate the presence of microorganisms on surfaces or in the environment, where such organisms can be deleterious to humans or the environment.
Specifications
Capacity (English)8.45 oz.
Closure ColorOrange
Closure MaterialPolypropylene
DescriptionTop style: Threaded
Diameter (Metric) Outer Neck3.1 cm
Capacity (Metric)250 mL
MaterialPolycarbonate
For Use With (Application)For shaker culture applications
Graduations25 mL
Neck StyleWide Mouth
No. per Case50/Cs.
No. per Pack1/Pk.
DisposableSingle-use
Mouth StyleWide
SterilitySterile
Closure TypeFlat
Unit SizePK