QuickFit™ Borosilicate Glass Test Tubes with Ground Socket
QuickFit™ Borosilicate Glass Test Tubes with Ground Socket
Actual product may vary
Fisherbrand
QuickFit™ Borosilicate Glass Test Tubes with Ground Socket
Manufactured from borosilicate glass for excellent chemical and thermal shock resistance. Quickfit™ Borosilicate Glass Test Tubes with Ground Socket are ideal for mixing and heating chemicals in a lab.
Have Questions?
Catalog NumberDiameter (Metric)Capacity (Metric)
FKS1071510141 mm195 mL
Catalog number FKS10715101
Price (SGD)
560.00
PK
Add to cart
Diameter (Metric):
41 mm
Capacity (Metric):
195 mL
Price (SGD)
560.00
PK
Add to cart
Manufactured from borosilicate glass for excellent chemical and thermal shock resistance. Quickfit™ Borosilicate Glass Test Tubes with Ground Socket are ideal for mixing and heating chemicals in a lab.
  • Presicion-ground socket ensures exact fit for stopper
  • Test tube sizes correspond with Quickfit™ Standard Taper Flat Head Borosilicate Glass Hollow Stoppers
  • Order tubes and stoppers separately
Specifications
Design TypeGround Socket
DescriptionStopper Size: 34/35
Capacity (Metric)195 mL
Length (Metric)250 mm
MaterialBorosilicate Glass
For Use WithQuickfit Hollow Stopper, Fisher Scientific product code 11349623
Quantity5 Pack
Diameter (Metric)41 mm
Unit SizePK