PELTOR™ Optime™ I Ear Muffs
PELTOR™ Optime™ I Ear Muffs
Actual product may vary
PELTOR™ Optime™ I Ear Muffs
PELTOR™ Optime™ I Ear Muffs
PELTOR™ Optime™ I Ear Muffs
PELTOR™ Optime™ I Ear Muffs
PELTOR™ Optime™ I Ear Muffs
PELTOR™ Optime™ I Ear Muffs
PELTOR™ Optime™ I Ear Muffs
PELTOR™ Optime™ I Ear Muffs
PELTOR™ Optime™ I Ear Muffs
PELTOR™ Optime™ I Ear Muffs
PELTOR™ Optime™ I Ear Muffs
PELTOR™ Optime™ I Ear Muffs
Fisherbrand
PELTOR™ Optime™ I Ear Muffs
PELTOR™ Optime™ I Ear Muffs are very lightweight, providing high wearer comfort. Ideal for protection against noise arising from a wide range of applications in the workplace and leisure activities. Offer versatile protection for both short and long duration tasks.
Have Questions?
Catalog NumberModelStyle
FKS10709535H510A-401-GUOver-the-head
Catalog number FKS10709535
Price (SGD)
-
Model:
H510A-401-GU
Style:
Over-the-head
  • Low-profile design which minimizes obtrusiveness
  • Large space inside cup helps reduce moisture and heat build-up
  • Easy-to-replace cushions and inserts helps keep them hygienically clean
  • Soft, wide cushions help reduce pressure around the ears and improve comfort and wearability
Specifications
ColorYellow
Certifications/ComplianceCE Approved
For Use With (Application)Provide Protection Against Noise Arising From a Wide Range of Applications in the Workplace and Leisure Activities
ModelH510A-401-GU
Noise Reduction Rating (NRR)27 dB
Plug ShapeOval
Product TypeI Ear Muffs
StyleOver-the-head
Unit SizeEA