Polypropylene Protein LoBind Microcentrifuge Tube
Polypropylene Protein LoBind Microcentrifuge Tube
Actual product may vary
Fisher Catalogue
Polypropylene Protein LoBind Microcentrifuge Tube
Store peptide, antibody or virus samples in these LoBind microcentrifuge tubes. Eppendorf™ Polypropylene Protein LoBind Microcentrifuge Tubes are ideal for work in protein research, mass spectrometry and protein array sample preparation.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)
FKS107087041.5 mL
AIT00301080940.5 mL
FKS103167520.5 mL
Catalog number FKS10708704
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Capacity (Metric):
1.5 mL
Store peptide, antibody or virus samples in these LoBind microcentrifuge tubes. Eppendorf™ Polypropylene Protein LoBind Microcentrifuge Tubes are ideal for work in protein research, mass spectrometry and protein array sample preparation.
  • Problem free handling with the tiniest volumes/concentrations of a variety of proteins
  • Preliminary handling of tubes by saturation with BSA or siliconisation is not required in order to guarantee the recovery of the proteins in the plastic tube
  • Recovery rates of more than 99% are achievable
  • Enzyme activities, for example, are not reduced by this tube, and denaturing effects caused by interaction with the surface are almost completely excluded
  • No undesired side effects of a surface coating
  • Tubes are available in Eppendorf LoBind™ material for maximum recovery of valuable samples
  • Maximum protein recovery (over 97%, BSA 1μg/mL)
  • Free from surface coating, e.g., silicone
  • Free from DNA, DNase, RNase and ATP (PCR* clean)
  • Special polypropylene material manufactured under optimized processing conditions
  • High degree of centrifugation durability, rated to 18, 000xg
  • High degree of transparency
  • Safe-Lock lid
Specifications
Closure MaterialPolypropylene
Capacity (Metric)1.5 mL
ColorTransparent
MaterialPolypropylene
For Use With (Application)Protein LoBind consumables are best suited for work in protein research, mass spectrometry, protein array sample preparation, and storage of peptide, antibody or virus samples
No. per Pack100 Tubes (2 bags of 50)
Volume (Metric)1.5mL
SterilityPCR Clean
Max. RCF18,000 x g
Unit SizePK