Stainless Steel Photographic Tray
Stainless Steel Photographic Tray
Actual product may vary
Fisherbrand
Stainless Steel Photographic Tray
Have Questions?
Catalog NumberLength (Metric)
FKS10708541190, 225 mm
Catalog number FKS10708541
Price (SGD)
418.00
EA
Add to cart
Length (Metric):
190, 225 mm
Price (SGD)
418.00
EA
Add to cart
Specifications
Dimensions (L x W x H)190 x 140 x 50 mm
Height (Metric)50 mm
Length (Metric)190, 225 mm
MaterialStainless Steel
TypeLaboratory Tray
Width (Metric)140, 175 mm
Quantity1 Each
ShapeRectangular
Unit SizeEA