Amicon™ Ultra-0.5 Centrifugal Filter Units
Amicon™ Ultra-0.5 Centrifugal Filter Units
Actual product may vary
Fisherbrand
Amicon™ Ultra-0.5 Centrifugal Filter Units
Achieve both high sample recovery and fast sample processing. EMD Amicon™ Ultra Centrifugal Filter Units feature regenerated cellulose membranes that are ideal for protein & nucleic acid purification, concentration and desalting.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberMWCOQuantity
FKS107046453 kDa96Pack
FISUFC510024100 kDa24Pack
Catalog number FKS10704645
Price (SGD)
2,523.00
PK
Add to cart
MWCO:
3 kDa
Quantity:
96Pack
Price (SGD)
2,523.00
PK
Add to cart
Achieve both high sample recovery and fast sample processing. EMD Amicon™ Ultra Centrifugal Filter Units feature regenerated cellulose membranes that are ideal for protein & nucleic acid purification, concentration and desalting.
  • High-recovery Ultracel™-PL regenerated cellulose membranes with MWCOs of 3, 10, 30, 50 or 100kDa
  • Vertical membrane reduces polarization for ultrafast spin times (as little as 10 to 15 min.)
  • Heat-sealed membrane eliminates possibility of downstream extractables
  • Universal rotor compatibility
  • High retentate recovery of >90%
  • High concentration factors of 80- to 100-fold (minimum final concentrate volume: 200μL)
  • Easy sample monitoring due to translucent housing and volume gradations
  • Direct pipetter sample access eliminates processing step to recover concentrate
Centrifugal filter enables
  • Concentration of biological samples containing antigens, antibodies, enzymes, nucleic acids, or microorganisms
  • Purification of macromolecular components found in tissue culture extracts or cell lysates
  • Concentration of dilute or prepurified proteins from column eluents
  • Desalting and buffer exchange
Specifications
DisposableSingle-use
Material (Membrane)High-recovery Ultracel-PL Regenerated Cellulose
MWCO3 kDa
SterilityNon-sterile
Filtration Area1 cm2
For Use With (Application)Protein purification, protein concentration and desalting
Housing MaterialRegenerated Cellulose (RC)
IncludesTwo microcentrifuge tubes
Max. Force14,000 xG for concentration spin; 1000 xG for reverse spin
Quantity96Pack
TypeCentrifugal Filters
Volume (Metric)0.5 mL
Length (English)0.196 in.
Length (Metric)4.99 cm
Diameter (Metric)10.8 mm
Unit SizePK