Ultrospec™ 10 Cell Density Meter measures density of cells in suspension at 600 nm/OD600.
Have Questions?
Catalog Number
Display Type
FKS10704417
LED
Catalog number FKS10704417
Price (SGD)
4,629.00
EA
Add to cart
Display Type:
LED
Price (SGD)
4,629.00
EA
Add to cart
Small, portable, and easy to use, the instrument can measure growth rates of E. coli, other bacteria, and yeast
Battery operated and portable for use where cells are actually cultured
Uses LED source in combination with an advanced fiber optic
Hand-held
Accepts tubes or plastic, disposable cells
Can be easily employed in incubation cabinets, under anaerobic conditions, and in other areas in the laboratory where cells are cultured
Spillages can be easily wiped from the smooth surface, and the cell compartment area can be flushed with formaldehyde or ethylene oxide to sterilize it
Download results to a PC or printer for convenient data storage and retrieval
Serial output with 99-place memory
Compact, low-maintenance device
Rechargeable batteries provide nearly one month of cord-free use