Check Order Status
Chroma Gesellschaft™ 13mm Plier Style Decapper
Fisherbrand
Chroma Gesellschaft™ 13mm Plier Style Decapper
Have Questions?
Catalog Number
dimClosureType
FKS10701822
Catalog number
FKS10701822
Price (SGD)
553.00
EA
Add to cart
Price (SGD)
553.00
EA
Add to cart
Specifications
Diameter (Metric) Cap
11 mm
Type
Plier-style Decapper
For Use With (Equipment)
11 mm Caps
Vial Type
Crimp Top Vial