Wheaton™ Glass Staining Dish for 10 or 20 Slides
Wheaton™ Glass Staining Dish for 10 or 20 Slides
Actual product may vary
Fisherbrand
Wheaton™ Glass Staining Dish for 10 or 20 Slides
Grooved staining dish holds up to 10 single slides or 20 slides back-to-back
Have Questions?
Catalog number FKS10693523
Price (SGD)
1,004.00
PK
Add to cart
Price (SGD)
1,004.00
PK
Add to cart
  • Immerse standard 3 × 1 in. (75 × 25mm) slides horizontally
  • Dish and cover made from soda lime glass
Specifications
Closure IncludedClosure Included
Closure MaterialGlass
Bottom ShapeRectangular
MaterialSoda Lime Glass
TypeStaining Dish
IncludesLid
Slide Count10 single slides, 19 slides arranged alternately straight across and diagonally, or 20 slides back-to-back of standard size
Dimensions (L x W)3.1 x 2.1 in. (8 x 5.5 cm)
Length (English)3.1 in.
Length (Metric)80 mm
Width (English)2.1 in.
Width (Metric)55 mm
Depth (English) Inside1.3 in.
Depth (Metric)35 mm
Depth (Metric) Inside3.5 cm
Unit SizePK