96-Well Cluster Tube System
96-Well Cluster Tube System
Actual product may vary
Fisherbrand
96-Well Cluster Tube System
Manufactured from polypropylene for strength and durability. Corning™ Cluster Tube System is ideal for harvesting cells, cell uptake and receptor binding assays, and radioimmunoassays.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberProduct TypeFormatQuantitySterility
FKS10690112Microplate Cluster TubeTube96/RackSterile
FIS07-200-317Microplate Cluster TubeTubeBulkNon-sterile
FIS07-200-319Microplate Cluster TubeTube96/RackNon-sterile
FIS07-200-320Microplate Cluster TubeTube96/RackSterile
Catalog number FKS10690112
Price (SGD)
1,051.00
PK
Add to cart
Product Type:
Microplate Cluster Tube
Format:
Tube
Quantity:
96/Rack
Sterility:
Sterile
Price (SGD)
1,051.00
PK
Add to cart
  • Composed of 96 polypropylene tubes in a standard microplate format.
  • Features 1.2 mL tubes that are available individually or in strips of eight tubes.
  • Polyethylene tube caps are available in 8-cap strips.
Specifications
FormatTube
Quantity96/Rack
AutoclavableAutoclavable
MaterialPolypropylene
Product TypeMicroplate Cluster Tube
SterilitySterile
No. per Pack96/Pk.
No. per Case960/Cs.
ColorTranslucent
Volume (Metric)1 mL
IncludesRack of 96 tubes. (Caps are sold separately.)
For Use With (Equipment)Standard 96-well microplate for harvesting cells, cell uptake and receptor binding assays and radioimmunoassays, useful with single and multichannel pipetters and in robotic workstations and centrifuges
Volume (English)0.033 oz.
Well Count96
Unit SizePK