Check Order Status
Filtration and Vaccum Dessiccators
Actual product may vary
Fisherbrand
Filtration and Vaccum Dessiccators
Can be evacuated to 1bar, with loss not exceeding 0.03mbar in 24 hours. Kartell™ Filtration and Vacuum Dessicators is implosion proof.
Have Questions?
Catalog Number
Height (Metric) Interior
Height (Metric)
Volume (Metric)
Diameter (Metric) Outer
FKS10683272
175 mm
230 mm
4.35 L
200 mm
Catalog number
FKS10683272
Price (SGD)
393.00
EA
Add to cart
Height (Metric) Interior:
175 mm
Height (Metric):
230 mm
Volume (Metric):
4.35 L
Diameter (Metric) Outer:
200 mm
Price (SGD)
393.00
EA
Add to cart
Transparent polycarbonate cover seals onto a neoprene O-ring set into the flange of the polypropylene base
Initial vacuum established with a little grease spread on the O-ring
Removable pan for holding a drying agent
Includes vacuum release tap/non-return valve assembly
Specially grooved stopper in assembly allows air, when required, to re-enter very slowly to avoid disturbing the contents
Specifications
Material
Polypropylene, Polycarbonate
Quantity
1 Each
Volume (Metric)
4.35 L
Height (Metric)
230 mm
Height (Metric) Interior
175 mm
Diameter (Metric) Outer
200 mm