Polypropylene Urine Containers
Polypropylene Urine Containers
Actual product may vary
Fisher Catalogue
Polypropylene Urine Containers
Manufactured from clear polypropylene for strength and chemical resistance. Kartell™ Polypropylene Urine Containers are available in sterile and non-sterile with color-coded screw caps.
Have Questions?
Catalog NumberSterilityClosure ColorNo. per Pack
FKS10682892SterileRed250/Pk.
Catalog number FKS10682892
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Sterility:
Sterile
Closure Color:
Red
No. per Pack:
250/Pk.
  • Non sterile cups have blue screw caps; sold in multi-packs
  • Sterile cups have red screw cap and a sanitized marking area; individually wrapped
  • CE marked; IVD approved
Specifications
Closure IncludedYes
GraduatedYes
MaterialPolypropylene
SterilitySterile
Capacity (Metric)150 mL
Certifications/ComplianceCE and IVD
Closure ColorRed
Closure MaterialPolypropylene
Closure TypeScrew Cap
Diameter (Metric)63 mm
For Use With (Application)Urine collection
Graduations20 to 100 mL
Height (Metric)73 mm
IncludesPolypropylene Screw Cap
No. per Pack250/Pk.
Unit SizePK