Racks for Matrix™ 2D Barcoded Storage Tubes
Racks for Matrix™ 2D Barcoded Storage Tubes
Actual product may vary
Racks for Matrix™ 2D Barcoded Storage Tubes
Racks for Matrix™ 2D Barcoded Storage Tubes
Racks for Matrix™ 2D Barcoded Storage Tubes
Racks for Matrix™ 2D Barcoded Storage Tubes
Fisherbrand
Racks for Matrix™ 2D Barcoded Storage Tubes
Create custom arrays with these empty ANSI-format tube storage racks with lid. Compatible with automated systems, rack features locking lid to secure samples.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberProduct TypeFor Use With (Equipment)
FKS10680214Barcoded Latch RackScrewtop Tubes (1.0 mL)
4896Barcoded Latch RackTubes (0.75 mL)
4897Barcoded Latch RackScrewtop Tubes (1.0 mL)
4900Recirculating ChillerCup horn or microplate horn of Q500/Q700 Sonicator™, Cup horn or microplate horn of Q500/Q700 Sonicator™
Catalog number FKS10680214
Price (SGD)
347.00
PK
Add to cart
Product Type:
Barcoded Latch Rack
For Use With (Equipment):
Screwtop Tubes (1.0 mL)
Price (SGD)
347.00
PK
Add to cart
Create custom arrays with these empty ANSI-format Racks for Thermo Scientific™ Matrix™ 2D Barcoded Storage Tubes. Racks are available in a proprietary, specially designed, stackable microplate format and feature locking lids to secure samples.
Superior Storage Rack Design
  • Unlike traditional storage tubes or blocks, Matrix 96-format 2D barcoded storage tubes are available in a proprietary, specially designed, stackable, microplate footprint
Latch Racks to Save Precious Space
  • The latch rack design enables manual multichannel pipette access to 2D tubes and eliminates contamination risk with lid design that does not contact the benchtop
  • The latch rack lid can be lifted robotically for 2D tube access with automated liquid handling systems and high throughput applications
Snap Rack - For users of traditional polycarbonate racks, Matrix tubes are also available in the Snap Rack. The Snap Rack has an open-bottom design for reading alphanumeric or TrakMates tubes, and is designed to hold samples securely during low-temperature storage and shipping.
Specifications
ColorWhite
Product TypeBarcoded Latch Rack
Capacity (Metric)1 mL
MaterialPlastic
For Use With (Equipment)Screwtop Tubes (1.0 mL)
Holds2D Matrix Tubes
Packaging Quantity5 Case
Capacity (English)0.0338 fl oz.
Unit SizePK