Stripette™ All-Plastic Wrapped, Polystyrene Serological Pipets, Sterile
Stripette™ All-Plastic Wrapped, Polystyrene Serological Pipets, Sterile
Actual product may vary
Fisherbrand
Stripette™ All-Plastic Wrapped, Polystyrene Serological Pipets, Sterile
Individually wrapped in easy-to-open plastic wrap for aseptic removal and elimination of static cling when wearing latex gloves
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)GraduationsColor
FKS1067734110 mL0.1 mLOrange
FIS07-200-1750 mL0.5 mLPurple
FIS07-200-72 mL0.01 mLGreen
FIS07-200-95 mL0.1 mLBlue
FKS101274005 mL0.1 mLBlue
FIS07-200-1210 mL0.1 mLOrange
FIS07-200-1525 mL0.2 mLRed
FIS07-200-547100 mL1 mLBlue
Catalog number FKS10677341
Price (SGD)
304.00
PK
Add to cart
Volume (Metric):
10 mL
Graduations:
0.1 mL
Color:
Orange
Price (SGD)
304.00
PK
Add to cart
Features an easy-opening plastic wrap for aseptic removal and elimination of static cling when wearing latex gloves. Corning™ Stripette™ Disposable Plastic-Wrapped Polystyrene Serological Pipets include a unique color-coded magnifying stripe for easier meniscus viewing.
  • Sterile
  • RNase-/DNase-free
  • Nonpyrogenic
  • Noncytotoxic
  • BSE/TSE-free
  • Exclusive anti-drip tip assures accurate delivery
  • Color-coded magnifier stripes make volume reading and size selection easier
  • Bidirectional graduations provide choice of ascending and descending scales
  • Negative graduations allow additional working volume
  • Three packaging options: Individually wrapped, paper/plastic; Individually wrapped, clear plastic; bulk packed
  • Volumetric accuracy of ±2% at full capacity
Specifications
Accuracy±2%
Certifications/ComplianceNon-pyrogenic, DNase, RNase-free
MaterialPolystyrene
SterilitySterile
GraduatedYes
Graduations0.1 mL
Length (English)13.54 in.
Negative Graduations3 mL
Tip StyleRound
Packaging TypeIndividually Wrapped, Plastic
ColorOrange
Volume (Metric)10 mL
Length (Metric)34.4 cm
TypeSerological Pipet