Nalgene™ Polypropylene Autoclaving Baskets
Nalgene™ Polypropylene Autoclaving Baskets
Actual product may vary
Nalgene™ Polypropylene Autoclaving Baskets
Nalgene™ Polypropylene Autoclaving Baskets
Nalgene™ Polypropylene Autoclaving Baskets
Fisher Catalogue
Nalgene™ Polypropylene Autoclaving Baskets
Hold labware during autoclaving and washing with these polypropylene autoclaving baskets, which also allow for complete venting and draining.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberHeight (Metric)Length (Metric)Width (Metric)
FKS1067654110.4 cm15.6 cm10.4 cm
6917-0127
also known as 6917-0127A
10.4 cm15.6 cm10.4 cm
6917-015017.8 cm15.6 cm17.8 cm
FKS1025786017.8 cm15.6 cm17.8 cm
Catalog number FKS10676541
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Height (Metric):
10.4 cm
Length (Metric):
15.6 cm
Width (Metric):
10.4 cm
Hold labware during autoclaving and washing with Thermo Scientific™ Nalgene™ Polypropylene Autoclaving Baskets, which also allow for complete venting and draining. These baskets won't corrode and are chemically compatible with most acids, bases and alcohols. The smooth surface will not scratch plastic or glass contents.
  • Nestable for storage
  • Autoclavable
Recycling information:
Please recycle the corrugate cartons and any LDPE bags your Nalgene products are delivered in.
Specifications
Product TypePolypropylene Autoclaving Basket
DescriptionAutoclaving Basket PP 105 x 123 x 154mm
AutoclavableAutoclavable
ColorWhite
MaterialPolypropylene
ShapeSquare
No. per Case6
No. per Pack1
Dimensions (L x W x H)6.125 x 4.87 x 4.125 in. (15.6 x 10.4 x 10.4cm)
Length (Metric)15.6 cm
Length (English)6.125 in.
Width (Metric)10.4 cm
Width (English)4.87 in.
Height (Metric)10.4 cm
Height (English)4.125 in.
Unit SizeEA