Elma Clean 65 Cleaning Concentrate
Product Image
Fisherbrand
Elma Clean 65 Cleaning Concentrate
Suitable for all cleaning applications which require very gentle treatment of the items to be cleaned. Fisherbrand™ Elma Clean 65 Cleaning Concentrate has been developed particularly for use in ultrasonic baths.
Have Questions?
Catalog NumberVolume (Metric)
FKS106753011 L
Catalog number FKS10675301
Price (SGD)
74.00
EA
Add to cart
Volume (Metric):
1 L
Price (SGD)
74.00
EA
Add to cart
  • Use for delicate laboratory instruments and equipment
  • Excellent for sensitive materials such as glass, plastics, metals and rubber
  • Removes contamination such as dust, fingerprints, pigments and light greases/oils
Specifications
FormLiquid
pH7
AppearanceYellow
Boiling Point≥100°C
CompositionAnionic surfactants 5 to 15%, non-ionic surfactants 15 to 30%, Amino-soaps <5%, NTA <5%, Phosp
ConcentrationUltrasonic bath 2 to 5%
Container TypeBottle
Volume (Metric)1 L
Unit SizeEA
Contents & Storage
-5°C to 30°C