Silicone Rubber Hand Protector Heat Resistant Glove
Silicone Rubber Hand Protector Heat Resistant Glove
Actual product may vary
Fisherbrand
Silicone Rubber Hand Protector Heat Resistant Glove
Protect your hands while handling hot labware with this glove. Saint Gobain Life Sciences Silicone Rubber Hand Protector Heat Resistant Glove features multiple concave tipped studs for a non-slip grip.
Have Questions?
Catalog number FKS10672571
Price (SGD)
137.00
EA
Add to cart
Price (SGD)
137.00
EA
Add to cart
  • One size
  • Manufactured from strong tear resistant silicone rubber
  • Withstands temperatures as high as 260°C and as low as -57°C
  • CE marked
Specifications
Laboratory/Cleanroom UseYes
MaterialSilicone Rubber
PowderedPowder Free
Product TypeGloves
Temperature (Metric) Max260°C
Quantity1 Each
Resistant ToHeat
Length (Metric)190 mm
Width (Metric)98 mm
Unit SizeEA