Nalgene™ Single Use Analytical Filter Funnels
Nalgene™ Single Use Analytical Filter Funnels
Actual product may vary
Nalgene™ Single Use Analytical Filter Funnels
Nalgene™ Single Use Analytical Filter Funnels
Nalgene™ Single Use Analytical Filter Funnels
Nalgene™ Single Use Analytical Filter Funnels
Fisherbrand
Nalgene™ Single Use Analytical Filter Funnels
Perform microbiological QC testing and analysis of water, food/beverage, raw material and finished product with these economical sterile disposable funnels.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Pore SizeColor
FKS10664661100 mL0.45 μmWhite with Black Grid
FIS09-740-30D100 mL0.45 μmWhite with Black Grid
FIS09-740-30K250 mL0.45 μmWhite with Black Grid
FKS10335801250 mL0.45 μmWhite with Black Grid
FIS12-456-666250 mL0.45 μmGray with Black Grid
Catalog number FKS10664661
Price (SGD)
989.00
PK
Add to cart
Capacity (Metric):
100 mL
Pore Size:
0.45 μm
Color:
White with Black Grid
Price (SGD)
989.00
PK
Add to cart
Use these economical sterile disposable filter funnels to perform microbiological QC testing and analysis of water, food/beverage, raw material and finished product. The squeeze and twist funnel is easy to remove for retrieval of the CN membrane.
Features:
  • High impact polystyrene collars are rugged for reliable performance
  • Preassembled, sterile, individually bagged to ensure sample integrity
  • Adapters accommodate No. 8 stoppers for use with 0.5 in. hole in vacuum manifolds or filter flasks
  • 47mm diameter cellulose nitrate membrane is certified for water quality work
  • 250mL sizes are useful when testing beverages that may foam
  • Cat. No. 147-0045 features gray membrane that turns black when wet, making it easier to detect and count growing colonies
Note: Designed for single use only
Includes: Six funnel adapters with each case
Specifications
Capacity (Metric)100 mL
Certifications/ComplianceMeet requirements of APHA Standard Methods and EPA requirements for water quality work.
ColorWhite with Black Grid
Diameter (Metric) Filter47 mm
For Use With (Equipment)Filter Manifold, Filtering Flask
Pore Size0.45 μm
Quantity50 Pk.
Product TypeFiltering Funnel
Material (Membrane)Cellulose Nitrate
Unit SizePK