Search Thermo Fisher Scientific
Catalog Number | Capacity (Metric) | Closure Size |
---|---|---|
FKS10664212 | 250 mL | 43 mm |
2189-0001 also known as 2189-0001/PF | 30 mL | 28 mm |
FIS03-313-4A | 30 mL | 28 mm |
FKS10214071 | 30 mL | 28 mm |
2189-0002 also known as 2189-0002/PF | 60 mL | 28 mm |
2189-0004 also known as 2189-0004/PF | 125 mL | 38 mm |
FIS03-313-4C | 125 mL | 38 mm |
2189-0008 also known as 2189-0008/PF | 250 mL | 43 mm |
FIS03-313-4D | 250 mL | 43 mm |
2189-0016 also known as 2189-0016/PF | 500 mL | 53 mm |
2189-0032 | 1 L | 63 mm |
FIS03-313-4F | 1 L | 63 mm |
FKS10505021 | 1 L | 63 mm |
Perform general purpose laboratory applications with Thermo Scientific™ Nalgene™ Wide-Mouth HDPE Economy Bottles with Closure. These bottles are a lighter-weight and more flexible version of Nalgene Lab Quality Bottles and are leakproof† when used with Nalgene closures. Made from high-quality laboratory-grade plastic materials for reliably low leachables and extractables. This product is ACT Labeled by My Green Lab to enable data-based sustainable product purchasing decisions.
† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.