Manufactured with optical polystyrene or UV grade PMMA for clear and sharp optical path with indication of optical path direction. Kartell™ Spectrophometry Cuvettes are ideal for spectrophotometry application.
Have Questions?
Change view
Catalog Number
Capacity (Metric)
Wavelength Range
Type
FKS10663852
1.5 mL
280 to 800 nm
Semi-micro Cell
FKS10462511
4.5 mL
280 to 800 nm
Macro Cell
2 Options
Catalog number FKS10663852
Price (SGD)
69.00
PK
Estimated availability date 01-Oct-2025
Add to cart
Capacity (Metric):
1.5 mL
Wavelength Range:
280 to 800 nm
Type:
Semi-micro Cell
Price (SGD)
69.00
PK
Add to cart
Manufactured with optical polystyrene or UV grade PMMA for clear and sharp optical path with indication of optical path direction. Kartell™ Spectrophometry Cuvettes are ideal for spectrophotometry application.
Equipped with optically machined walls (optical faces) to ensure optimal transparency along the spectral field
Polystrene range:340nm to 800nm
UV range: 280 to 800nm
Path length 10mm
Compatible with most spectrophotometers and photometers on the market