Translucent Polypropylene Tall Form Measuring Cylinders with Blue Graduations
Translucent Polypropylene Tall Form Measuring Cylinders with Blue Graduations
Actual product may vary
Fisherbrand
Translucent Polypropylene Tall Form Measuring Cylinders with Blue Graduations
Manufactured from highly translucent polypropylene for durability, clarity and good chemical resistance. Autoclavable Kartell™ Polypropylene Tall Form Measuring Cylinders with Blue Graduations provide an economical and safe solution for measuring the volume of liquids in a laboratory.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Graduations
FKS106632821000 mL100 mL
FKS10653282500 mL50 mL
Catalog number FKS10663282
Price (SGD)
67.00
EA
Add to cart
Capacity (Metric):
1000 mL
Graduations:
100 mL
Price (SGD)
67.00
EA
Add to cart
  • Permanent enhanced pin sharp blue graduations provide clarity and reproducibility
  • Polypropylene construction withstands temperatures up to 120°C 
  • Tall form, pentagon base for stability
  • Class B
  • Autoclavable
  • Suitable for foodstuff
Specifications
Capacity (Metric)1000 mL
MaterialPolypropylene
Tolerance±10 mL
Certifications/ComplianceISO 6706-1981 (E)
ClassB
ScalePermanent blue graduations
Diameter (Metric)66 mm
Graduations100 mL
AutoclavableAutoclavable
Limit of Error±10mL
Max. Temperature100°C
Subdivision10mL
ShapeCylindrical
For Use With (Application)Foodstuff
Scale ColorBlue
Height (Metric)439 mm
TypeGraduated Measuring Cylinder
Unit SizeEA