Check Order Status
Basic Line Conductivity Standard, 1.3μS/cm, Hamilton™
Structure Search
Fisherbrand
Basic Line Conductivity Standard, 1.3μS/cm, Hamilton™
Provides unprecedented stability. Hamilton™ Basic Line Conductivity Standard, 1.3μS/cm comes with a certificate with calibration document form DFM.
Have Questions?
Catalog Number
Quantity
FKS10661794
250 mL
Catalog number
FKS10661794
Price (SGD)
670.00
EA
Add to cart
Quantity:
250 mL
Request bulk or custom format
Price (SGD)
670.00
EA
Add to cart
Specifications
Packaging
Glass bottle
Quantity
250 mL
Unit Size
EA
Remains stable for up to 1 year
Expiration date shown on every bottle
Bottles are permitted to stay open for a total of 60 minutes