Volac™ Glass Pasteur Pipets
Volac™ Glass Pasteur Pipets
Actual product may vary
Fisherbrand
Volac™ Glass Pasteur Pipets
Use these pipets for a variety of applications. Poulten Graf™ Volac™ Glass Pasteur Pipets feature a slightly heavier wall thickness to provide greater strength and reduce the risk of breakage in use or when fitting PVC teats.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberLength (Metric)SterilityIncludes
FKS10659932150 mmNon-sterilePlugged
FKS10026041150 mmNon-sterileUnplugged
FKS10006021150 mmUnplugged
FKS10209381230 mmNon-sterileUnplugged
Catalog number FKS10659932
Price (SGD)
649.00
PK
Add to cart
Length (Metric):
150 mm
Sterility:
Non-sterile
Includes:
Plugged
Price (SGD)
649.00
PK
Add to cart
  • Available with or without plug
  • Glass pipets come in 150, 230 and 270mm lengths
  • Packing materials are carefully chosen to ensure ease of use for the technician with maximum protection from breakage during transit and minimum lint or dust contamination
  • All standard pipettes can be supplied with 'sealed points' and break mark
Specifications
Certifications/ComplianceBS5732,ISO7712
MaterialGlass
SterilityNon-sterile
Drops/mL30 to 35 mL
IncludesPlugged
Quantity1000 Pack
Length (Metric)150 mm
TypePasteur Pipet