Orion™ High Performance Ammonia Combination Ion Selective Electrode
Orion™ High Performance Ammonia Combination Ion Selective Electrode
Actual product may vary
Orion™ High Performance Ammonia Combination Ion Selective Electrode
Orion™ High Performance Ammonia Combination Ion Selective Electrode
Orion™ High Performance Ammonia Combination Ion Selective Electrode
Orion™ High Performance Ammonia Combination Ion Selective Electrode
Fisherbrand
Orion™ High Performance Ammonia Combination Ion Selective Electrode
Obtain quick, simple, accurate and economical results at low level ammonia measurements.
Have Questions?
Catalog NumberConnector Types
FKS10659663BNC (Waterproof)
Catalog number FKS10659663
Price (SGD)
3,783.00
EA
Add to cart
Connector Types:
BNC (Waterproof)
Price (SGD)
3,783.00
EA
Add to cart
Obtain a more linear response at lower levels and improved response time in ammonia measurements with the Thermo Scientific™ Orion™ High-Performance Ammonia Electrode, an ammonia gas-sensing electrode.
Ammonia is an important aspect of wastewater and environmental management. The 9512HPBNWP complies with SM 4500-NH3 D-1997 or E-1997; ASTM D1426-08 (B).
Features:
  • 9512HPBNWP features a hydrophobic gas-permeable membrane technology for use with either loose membrane 951214 or pre-assembled membrane caps 951215. Good for samples with <0.1 mg/L ammonia.
Solutions required:
  • Standards
    • 951006 0.1 M NH4Cl standard, 475 mL bottle
    • 951007 1000 ppm ammonia as nitrogen standard, 475 mL bottle
    • 951207 100 ppm ammonia as nitrogen standard, 475 mL bottle
  • ISA
    • 951011 Alkaline reagent, 475 ml bottle
    • 951210 Ammonia low level ISA with blue dye, 475 mL bottle
    • 951211 Ammonia ISA with blue dye, 475 mL bottle
  • Electrode fill solution
    • 951209 Electrode filling solution, 60 mL bottle
  • Storage solution
    • 951213 Ammonia electrode storage solution, 475 mL bottle
Consumables required:
  • Membranes
    • 951214 Loose membranes for HP electrodes, box of 20
    • 951215 Pre-assembled outer body and membrane cap assembly for HP electrodes, 3 pack
  • Items included with
    • 951209 Electrode filling solution, 60mL 2 bottles
    • 951214 Loose membranes, box of 20. 1 box
    • 951215 Pre-assembled outer body and membrane cap assembly for HP electrodes, 3 pack
Warranty period:
  • 12 months from date of purchase
Specifications
Connector TypesBNC (Waterproof)
DescriptionAmmonia High Performance Ion Selective Electrode
Diameter (Metric) Body12mm
Filling Solution TypeHigh performance ammonia electrode filling solution
Internal ReferenceVolatile amines
Length (Metric)120mm
Length (Metric) Cable1M
MaterialEpoxy
Temperature (English) Operating32° to 122°F
Temperature (Metric) Operating0°C to 50°C
TypeAmmonia Electrode
Unit SizeEA