FR-902 FFKM / PEEK / Stainless Steel Flow Restrictor
FR-902 FFKM / PEEK / Stainless Steel Flow Restrictor
Actual product may vary
Fisherbrand
FR-902 FFKM / PEEK / Stainless Steel Flow Restrictor
Used when using low pressure columns and/or when the pH electrode is mounted in the pH flow cell. Cytiva FR-902 FFKM / PEEK / Stainless Steel Flow Restrictor is pre-set to 0.2 MPa.
Have Questions?
Catalog NumberMaterial
FKS10658629FFKM / PEEK / Stainless Steel
Catalog number FKS10658629
Price (SGD)
3,102.00
EA
Add to cart
Material:
FFKM / PEEK / Stainless Steel
Price (SGD)
3,102.00
EA
Add to cart
  • The wetted parts are resistant to organic solvents and salt buffers commonly used in chromatography of biomolecules, except 100 % Ethyl acetate, 100 % Hexane and 100 % Tetrahydrofuran Complete
  • FFKM, Fully fluorinated perfluoroelastomer PEEK, Polyetheretherketone Stainless steel 
Specifications
TypeFlow Restrictor
MaterialFFKM / PEEK / Stainless Steel
Weight (Metric)0.1 kg
Unit SizeEA