LDPE Pasteur Pipettes
LDPE Pasteur Pipettes
Actual product may vary
Fisherbrand
LDPE Pasteur Pipettes
LDPE Pasteur Pipettes are ideal for aliquots with very good reproducibility of the number of drops per milliliter.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberLength (Metric)Volume (Metric)Diameter (Metric) OuterDrops/mL
FKS10646341152 mm2 mL3.3 mm22 to 26
FKS10100531152 mm3 mL2.8 mm25 to 27
Catalog number FKS10646341
Price (SGD)
241.00
PK
Add to cart
Length (Metric):
152 mm
Volume (Metric):
2 mL
Diameter (Metric) Outer:
3.3 mm
Drops/mL:
22 to 26
Price (SGD)
241.00
PK
Add to cart
  • Manufactured from LDPE.
  • Very good reproducibility of the number of drops per milliliter.
  • Ideal for aliquots.
  • Pasteur pipettes can be filled and deep-frozen, or changed into a closed vessel by heat-sealing the tip.
  • The integrated pipetting bulb depresses easily, minimizing fatigue from frequent pipetting.
  • Resistant to gas or gamma radiation sterilization.
Specifications
Diameter (Metric) Outer3.3 mm
MaterialLDPE
Drops/mL22 to 26
Quantity500 Pack
Top StyleBulb Shaped
Volume (Metric)2 mL
Length (Metric)152 mm
TypePasteur Pipet